Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FRMPD2
Entrez Gene ID
423777
Gene Name
FERM and PDZ domain containing 2
Gene Aliases
-
Chromosome Location
Chr.6: 18988952 - 19042427 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015288430.2 XP_015143916.1 22 - 23 3068 99
XM_025151960.1 XP_025007728.1 21 - 22 2960 99
XM_025151961.1 XP_025007729.1 22 - 23 3068 99
XM_025151962.1 XP_025007730.1 6 - 7 783 99
GenBank mRNA CR407180.1 - 4 - 5 585 99

View Full Product Details