Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MRPS35
Entrez Gene ID
419048
Gene Name
mitochondrial ribosomal protein S35
Gene Aliases
-
Chromosome Location
Chr.1: 72622760 - 72644857 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278091.1 NP_001265020.1 7 - 8 824 81
NM_001278092.1 NP_001265021.1 7 - 8 766 81
GenBank mRNA BX935160.1 - 7 - 8 758 81
CR522886.1 - 7 - 8 656 81

View Full Product Details