Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HOXD4
Entrez Gene ID
424139
Gene Name
homeobox D4
Gene Aliases
Chox-a
Chromosome Location
Chr.7: 16275972 - 16277619 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012293.1 NP_001012293.1 1 - 2 412 73
GenBank mRNA X52672.1 - - 378 73

View Full Product Details