Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SNRPB2
Entrez Gene ID
421261
Gene Name
small nuclear ribonucleoprotein polypeptide B2
Gene Aliases
-
Chromosome Location
Chr.3: 5652599 - 5658851 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_419331.6 XP_419331.1 4 - 5 500 107
GenBank mRNA BX931436.2 - 4 - 5 481 107
BX932844.2 - 4 - 5 482 107
BX933982.1 - 4 - 5 491 107
BX934618.1 - 4 - 5 494 107
BX935753.1 - 4 - 5 488 107

Gene Symbol
CTAGE1
Entrez Gene ID
395484
Gene Name
cutaneous T-cell lymphoma-associated antigen 1
Gene Aliases
OTOR
Chromosome Location
Chr.3: 5657272 - 5667265 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_025148519.1 XP_025004287.1 1 - 2 53 107

View Full Product Details