Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM268
Entrez Gene ID
417248
Gene Name
transmembrane protein 268
Gene Aliases
C17H9ORF91, C9orf91
Chromosome Location
Chr.17: 2882256 - 2891735 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004945796.3 XP_004945853.2 6 - 7 915 139
XM_015279690.2 XP_015135176.2 6 - 7 1067 139
XM_025141524.1 XP_024997292.1 6 - 7 1072 139
XM_415524.6 XP_415524.4 6 - 7 920 139
GenBank mRNA CR353484.1 - 6 - 7 930 139

View Full Product Details