Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FHL5
Entrez Gene ID
421803
Gene Name
four and a half LIM domains 5
Gene Aliases
-
Chromosome Location
Chr.3: 72780192 - 72808111 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277723.1 NP_001264652.1 2 - 3 155 73
GenBank mRNA BX929623.2 - 2 - 3 174 73

View Full Product Details