Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MTNR1B
Entrez Gene ID
396338
Gene Name
melatonin receptor 1B
Gene Aliases
Mel-1B-R, Mel1b
Chromosome Location
Chr.1: 187524139 - 187544369 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001293103.1 NP_001280032.1 2 - 2 966 122
GenBank mRNA U30609.1 - 1 - 1 750 122

View Full Product Details