Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CARD19
Entrez Gene ID
415987
Gene Name
caspase recruitment domain family member 19
Gene Aliases
C12H9ORF89, C9orf89
Chromosome Location
Chr.12: 7033642 - 7054153 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001232398.5 XP_001232399.1 3 - 4 339 66
XM_004944551.3 XP_004944608.1 3 - 4 429 66
XM_015293413.2 XP_015148899.1 3 - 4 448 66
XM_015293414.2 XP_015148900.1 3 - 4 321 66
XM_015293415.2 XP_015148901.1 3 - 4 509 66
XM_414329.5 XP_414329.1 3 - 4 429 66
GenBank mRNA BX935395.1 - 3 - 4 356 66
CR353336.1 - 1 - 2 719 66

View Full Product Details