Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LNP1
Entrez Gene ID
418388
Gene Name
leukemia NUP98 fusion partner 1
Gene Aliases
-
Chromosome Location
Chr.1: 86009787 - 86023742 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004938250.3 XP_004938307.1 3 - 4 435 80
XM_025143930.1 XP_024999698.1 3 - 4 436 80
XM_025143936.1 XP_024999704.1 3 - 4 431 80
XM_429574.6 XP_429574.1 3 - 4 415 80
GenBank mRNA CR352915.1 - 1 - 2 256 80

View Full Product Details