Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HS3ST1
Entrez Gene ID
422840
Gene Name
heparan sulfate-glucosamine 3-sulfotransferase 1
Gene Aliases
-
Chromosome Location
Chr.4: 78033667 - 78044779 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_420786.6 XP_420786.1 2 - 2 1238 108
GenBank mRNA BX931398.2 - 1 - 1 452 108
BX934256.2 - 1 - 1 983 108

View Full Product Details