Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TAF1D
Entrez Gene ID
419002
Gene Name
TATA-box binding protein associated factor, RNA polymerase I subunit D
Gene Aliases
JOSD3, OACA2, OACA3, SNORA32
Chromosome Location
Chr.1: 187114697 - 187125651 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015280465.2 XP_015135951.1 11 - 12 1511 141
XM_025146897.1 XP_025002665.1 12 - 13 1660 141
XM_417196.6 XP_417196.1 11 - 12 1555 141
GenBank mRNA BX929784.2 - 9 - 10 954 141

View Full Product Details