Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CFAP221
Entrez Gene ID
424272
Gene Name
cilia and flagella associated protein 221
Gene Aliases
PCDP1
Chromosome Location
Chr.7: 28330064 - 28350684 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004942966.3 XP_004943023.1 14 - 15 1468 102
XM_422119.6 XP_422119.4 13 - 14 1443 102
GenBank mRNA BX950354.2 - 4 - 5 384 102

View Full Product Details