Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NKX6-2
Entrez Gene ID
395591
Gene Name
NK6 homeobox 2
Gene Aliases
NKX-6.2
Chromosome Location
Chr.6: 36216483 - 36218896 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199548.1 NP_001186477.1 3 - 3 638 76
XM_015288987.1 XP_015144473.1 3 - 3 936 76
GenBank mRNA AF189767.1 - 1 - 1 52 76

View Full Product Details