Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VWC2
Entrez Gene ID
420944
Gene Name
von Willebrand factor C domain containing 2
Gene Aliases
-
Chromosome Location
Chr.2: 80493366 - 80551992 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015282290.2 XP_015137776.1 3 - 4 1088 63
XM_015282291.2 XP_015137777.1 3 - 4 1016 63
XM_419028.6 XP_419028.3 3 - 4 1072 63
GenBank mRNA BX931689.1 - 2 - 3 494 63
CR523060.1 - 2 - 3 917 63

View Full Product Details