Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KANSL1L
Entrez Gene ID
424004
Gene Name
KAT8 regulatory NSL complex subunit 1-like
Gene Aliases
C2orf67, C7H2ORF67, MSL1v2
Chromosome Location
Chr.7: 2714120 - 2771622 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015289308.2 XP_015144794.1 8 - 9 2192 142
XM_015289310.2 XP_015144796.1 7 - 8 1997 142
XM_015289312.2 XP_015144798.1 8 - 9 2120 142
GenBank mRNA BX932155.1 - 5 - 6 695 142

View Full Product Details