Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SDE2
Entrez Gene ID
421318
Gene Name
SDE2 telomere maintenance homolog
Gene Aliases
C1orf55, C3H1ORF55, dJ671D7.1
Chromosome Location
Chr.3: 17260776 - 17268370 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015283919.2 XP_015139405.1 2 - 3 252 116
GenBank mRNA BX934015.1 - 2 - 3 234 116

View Full Product Details