Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EIF3E
Entrez Gene ID
420272
Gene Name
eukaryotic translation initiation factor 3 subunit E
Gene Aliases
EIF3S6
Chromosome Location
Chr.2: 132199296 - 132219765 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006349.1 NP_001006349.1 6 - 7 614 92
GenBank mRNA AJ719829.1 - 6 - 7 614 92

View Full Product Details