Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WFDC1
Entrez Gene ID
395238
Gene Name
WAP four-disulfide core domain 1
Gene Aliases
PS20
Chromosome Location
Chr.11: 16725683 - 16738297 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204554.1 NP_989885.1 5 - 6 840 92
GenBank mRNA AJ438290.1 - 5 - 6 840 92
BX930581.2 - 5 - 6 678 92
BX931497.1 - 5 - 6 691 92

View Full Product Details