Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FKBP1B
Entrez Gene ID
395254
Gene Name
FK506 binding protein 1B
Gene Aliases
FKBP12.6
Chromosome Location
Chr.3: 104141299 - 104187704 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204567.1 NP_989898.1 2 - 3 132 104
GenBank mRNA AB074887.1 - 2 - 3 132 104

View Full Product Details