Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
ENS-3
Entrez Gene ID
395341
Gene Name
pol-like protein ENS-3
Gene Aliases
-
Chromosome Location
Chr.2: 71610700 - 71616305 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204632.1 NP_989963.1 1 - 1 1351 107
GenBank mRNA AF329451.1 - 1 - 1 1351 107

View Full Product Details