Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PPP1R9B
Entrez Gene ID
395762
Gene Name
protein phosphatase 1, regulatory (inhibitor) subunit 9B
Gene Aliases
-
Chromosome Location
Chr.7: 17655687 - 17697085 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204923.3 NP_990254.3 5 - 6 1905 92
GenBank mRNA AF043493.1 - 3 - 4 344 92

View Full Product Details