Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NPPA
Entrez Gene ID
395765
Gene Name
natriuretic peptide A
Gene Aliases
ANF, ANP, NPPB
Chromosome Location
Chr.21: 5578197 - 5580816 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204925.1 NP_990256.1 1 - 2 179 76
GenBank mRNA X57702.1 - - 179 76

View Full Product Details