Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GABPB1
Entrez Gene ID
415453
Gene Name
GA binding protein transcription factor beta subunit 1
Gene Aliases
GABPB2
Chromosome Location
Chr.10: 11304518 - 11323916 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005816.1 NP_001005816.1 3 - 4 336 81
NM_001012622.1 NP_001012640.1 3 - 4 344 81
GenBank mRNA AB188512.1 - 3 - 4 344 81
AB188513.1 - 2 - 3 245 81
AJ719707.1 - 3 - 4 336 81

View Full Product Details