Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PHAX
Entrez Gene ID
427124
Gene Name
phosphorylated adaptor for RNA export
Gene Aliases
RNUXA
Chromosome Location
Chr.Z: 56385502 - 56394442 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006575.2 NP_001006575.1 1 - 2 153 124
GenBank mRNA AJ719604.1 - 1 - 2 153 124

View Full Product Details