Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
POMGNT2
Entrez Gene ID
428446
Gene Name
protein O-linked mannose N-acetylglucosaminyltransferase 2 (beta 1,4-)
Gene Aliases
AGO61, C2H3ORF39, C3orf39, GTDC2, MDDGA8
Chromosome Location
Chr.2: 41054999 - 41087066 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012294.1 NP_001012294.1 1 - 1 1653 80
XM_015281574.2 XP_015137060.1 2 - 2 1807 80
XM_015281575.2 XP_015137061.1 2 - 2 1972 80
XM_015281576.2 XP_015137062.1 2 - 2 1870 80
GenBank mRNA AJ868536.1 - 1 - 1 1653 80

View Full Product Details