Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TTPAL
Entrez Gene ID
419197
Gene Name
alpha tocopherol transfer protein like
Gene Aliases
C20orf121
Chromosome Location
Chr.20: 5403488 - 5411241 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030854.1 NP_001026025.1 2 - 3 496 120
XM_025142193.1 XP_024997961.1 2 - 3 474 120
XM_025142194.1 XP_024997962.1 2 - 3 573 120
GenBank mRNA AJ851795.1 - 2 - 3 496 120

View Full Product Details