Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FABP3
Entrez Gene ID
419557
Gene Name
fatty acid binding protein 3
Gene Aliases
FABP, H-FABP
Chromosome Location
Chr.23: 703869 - 706870 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030889.1 NP_001026060.1 3 - 4 346 60
GenBank mRNA BX935814.2 - 3 - 4 368 60
JX090310.1 - 3 - 4 346 60
KT165376.1 - 3 - 4 346 60

View Full Product Details