Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GHRL
Entrez Gene ID
408185
Gene Name
ghrelin/obestatin prepropeptide
Gene Aliases
ghrelin, preproghrelin
Chromosome Location
Chr.12: 19849376 - 19852075 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001131.1 NP_001001131.1 2 - 3 263 95
GenBank mRNA AB075215.1 - 2 - 3 256 95
AY299454.1 - 2 - 3 263 95
BX931092.2 - 2 - 3 241 95
BX933754.2 - 1 - 2 78 95

View Full Product Details