Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TENM2
Entrez Gene ID
373854
Gene Name
teneurin transmembrane protein 2
Gene Aliases
ODZ2, TEN2
Chromosome Location
Chr.13: 5569975 - 6177414 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003718.1 NP_001003718.1 7 - 8 1518 61
NM_204097.2 NP_989428.2 8 - 9 1602 61
XM_015293567.2 XP_015149053.1 10 - 11 1875 61
XM_015293568.2 XP_015149054.1 10 - 11 1875 61
XM_015293569.2 XP_015149055.1 10 - 11 1875 61
XM_015293571.2 XP_015149057.1 9 - 10 1791 61
XM_015293573.2 XP_015149059.1 9 - 10 1752 61
XM_015293576.2 XP_015149062.1 9 - 10 1752 61
XM_015293578.2 XP_015149064.1 7 - 8 1581 61
XM_015293579.2 XP_015149065.1 6 - 7 1495 61
XM_015293581.2 XP_015149067.1 9 - 10 1791 61
XM_025154773.1 XP_025010541.1 7 - 8 1518 61
XM_025154774.1 XP_025010542.1 7 - 8 1518 61
XM_025154775.1 XP_025010543.1 9 - 10 1665 61
XM_025154776.1 XP_025010544.1 6 - 7 1395 61
XM_025154777.1 XP_025010545.1 8 - 9 1581 61
XM_025154778.1 XP_025010546.1 7 - 8 10015 61
GenBank mRNA AJ245711.1 - 7 - 8 1518 61
AJ279031.1 - 8 - 9 1602 61

View Full Product Details