Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ST8SIA1
Entrez Gene ID
395860
Gene Name
ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 1
Gene Aliases
SIAL-T2
Chromosome Location
Chr.1: 66588236 - 66704165 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205008.1 NP_990339.1 4 - 5 545 76
GenBank mRNA U73176.1 - 4 - 5 545 76

View Full Product Details