Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMOD1
Entrez Gene ID
395883
Gene Name
tropomodulin 1
Gene Aliases
E-Tmod, tropomodulin-1
Chromosome Location
Chr.Z: 70994947 - 71015318 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205027.1 NP_990358.1 1 - 2 127 92
XM_015280589.2 XP_015136075.1 2 - 3 189 92
XM_015280591.2 XP_015136077.1 2 - 3 1071 92
GenBank mRNA AB052132.1 - 1 - 2 78 92
L36678.1 - 1 - 2 127 92

View Full Product Details