Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RPS4Y1
Entrez Gene ID
396001
Gene Name
ribosomal protein S4, Y-linked 1
Gene Aliases
RPS4, RPS4X
Chromosome Location
Chr.4: 1889359 - 1892696 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205108.1 NP_990439.1 1 - 2 91 86
GenBank mRNA BX930717.1 - 1 - 2 92 86
CR391662.1 - 1 - 2 95 86
L24368.1 - 1 - 2 91 86

View Full Product Details