Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
NUCKS1
Entrez Gene ID
404775
Gene Name
nuclear casein kinase and cyclin dependent kinase substrate 1
Gene Aliases
JC7, NUCKS
Chromosome Location
Chr.26: 2264178 - 2277527 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213581.2 NP_998746.1 - 988 134
GenBank mRNA AB049826.1 - - 988 134
AJ720297.1 - - 988 134

View Full Product Details