Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UFSP2
Entrez Gene ID
422542
Gene Name
UFM1 specific peptidase 2
Gene Aliases
C4orf20
Chromosome Location
Chr.4: 39244669 - 39256259 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031136.1 NP_001026307.1 6 - 7 705 135
XM_015276225.2 XP_015131711.1 6 - 7 727 135
XM_015276226.2 XP_015131712.1 6 - 7 724 135
GenBank mRNA AJ720831.1 - 6 - 7 705 135

View Full Product Details