Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DTWD2
Entrez Gene ID
427386
Gene Name
DTW domain containing 2
Gene Aliases
-
Chromosome Location
Chr.Z: 71964912 - 72039739 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031442.1 NP_001026613.1 2 - 3 251 96
GenBank mRNA AJ719497.1 - 2 - 3 251 96

View Full Product Details