Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC25A32
Entrez Gene ID
428377
Gene Name
solute carrier family 25 member 32
Gene Aliases
-
Chromosome Location
Chr.2: 129295115 - 129309963 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031506.1 NP_001026677.1 6 - 7 855 133
GenBank mRNA AJ720399.1 - 6 - 7 855 133

View Full Product Details