The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001031515.1 | NP_001026686.1 | 1 - 1 | 947 | 107 |
XM_015282469.2 | XP_015137955.1 | 3 - 3 | 1294 | 107 | |
XM_015282470.1 | XP_015137956.1 | 2 - 2 | 1198 | 107 | |
GenBank mRNA | KC874994.1 | - | 1 - 1 | 947 | 107 |
KC874995.1 | - | 1 - 1 | 947 | 107 |