Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIM41
Entrez Gene ID
417043
Gene Name
tripartite motif containing 41
Gene Aliases
-
Chromosome Location
Chr.16: 2519951 - 2525938 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030672.2 NP_001025843.1 1 - 2 273 149

View Full Product Details