Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SET
Entrez Gene ID
417210
Gene Name
SET nuclear proto-oncogene
Gene Aliases
-
Chromosome Location
Chr.17: 5579042 - 5588504 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030691.2 NP_001025862.2 4 - 5 662 133
GenBank mRNA AJ851581.1 - 4 - 5 399 133

View Full Product Details