Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SARAF
Entrez Gene ID
422624
Gene Name
store-operated calcium entry associated regulatory factor
Gene Aliases
TMEM66
Chromosome Location
Chr.4: 49230262 - 49237926 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031146.1 NP_001026317.1 5 - 6 885 92
GenBank mRNA AJ720284.1 - 5 - 6 885 92

View Full Product Details