Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VTG2
Entrez Gene ID
424533
Gene Name
vitellogenin 2
Gene Aliases
VIT2, vit, vitellogenin-2
Chromosome Location
Chr.8: 16869042 - 16889747 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031276.1 NP_001026447.1 8 - 9 1083 96

View Full Product Details