Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MORN4
Entrez Gene ID
425290
Gene Name
MORN repeat containing 4
Gene Aliases
C10orf83
Chromosome Location
Chr.6: 23273221 - 23275696 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031335.1 NP_001026506.1 3 - 4 282 64
GenBank mRNA AJ720278.1 - 3 - 4 282 64

View Full Product Details