Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MRPL28
Entrez Gene ID
426121
Gene Name
mitochondrial ribosomal protein L28
Gene Aliases
-
Chromosome Location
Chr.14: 12534748 - 12537899 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031359.2 NP_001026530.2 3 - 4 604 69
GenBank mRNA BU311325.1 - 3 - 4 321 69
BU311826.1 - 2 - 3 232 69
BU380572.1 - 3 - 4 604 69
CO419869.1 - 2 - 3 527 69

View Full Product Details