Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
STUB1
Entrez Gene ID
426918
Gene Name
STIP1 homology and U-box containing protein 1
Gene Aliases
-
Chromosome Location
Chr.14: 13543813 - 13547618 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031406.2 NP_001026577.1 6 - 7 907 120
XM_015294667.1 XP_015150153.1 5 - 6 845 120
GenBank mRNA AJ720999.1 - 6 - 7 907 120

View Full Product Details