Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HNRNPAB
Entrez Gene ID
396268
Gene Name
heterogeneous nuclear ribonucleoprotein A/B
Gene Aliases
HNRPAB
Chromosome Location
Chr.13: 14985178 - 15015906 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205328.5 NP_990659.1 4 - 5 753 115
XM_015293745.2 XP_015149231.1 5 - 6 3899 115
XM_015293746.2 XP_015149232.1 5 - 6 3899 115
XM_015293747.2 XP_015149233.1 5 - 6 3899 115
XM_015293748.2 XP_015149234.1 5 - 6 3899 115
XM_015293750.2 XP_015149236.1 5 - 6 3899 115
XM_015293751.2 XP_015149237.1 5 - 6 3899 115
XM_015293752.2 XP_015149238.1 5 - 6 3899 115
GenBank mRNA CR352392.1 - 5 - 6 651 115
X80340.1 - - 757 115

View Full Product Details