Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GHRH
Entrez Gene ID
419178
Gene Name
growth hormone releasing hormone
Gene Aliases
-
Chromosome Location
Chr.20: 5042193 - 5053986 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040464.1 NP_001035554.1 5 - 6 524 75
XM_015296359.2 XP_015151845.1 5 - 6 4389 75
XM_015296360.2 XP_015151846.1 6 - 7 871 75
GenBank mRNA BX929984.1 - 5 - 6 640 75
DQ465017.1 - 5 - 6 524 75
DQ465018.1 - 5 - 6 524 75

View Full Product Details