Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C4
Entrez Gene ID
426611
Gene Name
complement 4
Gene Aliases
C4-2
Chromosome Location
Chr.16: 2610038 - 2624392 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077233.1 NP_001070701.1 29 - 30 3828 93
GenBank mRNA DQ224386.1 - 29 - 30 3828 93

View Full Product Details