Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AP2M1
Entrez Gene ID
770246
Gene Name
adaptor related protein complex 2 mu 1 subunit
Gene Aliases
-
Chromosome Location
Chr.9: 15973948 - 15996719 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079494.1 NP_001072962.1 5 - 6 666 100
XM_015291442.2 XP_015146928.1 7 - 8 656 100
XM_015291443.2 XP_015146929.1 7 - 8 726 100
XM_015291444.2 XP_015146930.1 6 - 7 665 100
XM_015291445.2 XP_015146931.1 6 - 7 673 100
XM_015291446.2 XP_015146932.1 6 - 7 737 100
XM_015291447.2 XP_015146933.1 5 - 6 651 100
XM_025153417.1 XP_025009185.1 5 - 6 681 100
GenBank mRNA AJ719338.1 - 5 - 6 666 100
CR385218.1 - 5 - 6 619 100

View Full Product Details