Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
KIFC1
Entrez Gene ID
417037
Gene Name
kinesin family member C1
Gene Aliases
HSET, KNSL2
Chromosome Location
Chr.16: 2439128 - 2443841 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081698.1 NP_001075167.1 7 - 8 1182 79
GenBank mRNA AY332739.1 - 7 - 8 1182 79

View Full Product Details