Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GLI2
Entrez Gene ID
395956
Gene Name
GLI family zinc finger 2
Gene Aliases
GLI2/GLI4, GLI3
Chromosome Location
Chr.7: 25774748 - 25958144 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271901.1 NP_001258830.1 6 - 7 937 92
NM_001271902.1 NP_001258831.1 6 - 7 834 92
XM_015289786.2 XP_015145272.1 6 - 7 1152 92
XM_015289787.2 XP_015145273.1 6 - 7 1153 92
XM_025152142.1 XP_025007910.1 7 - 8 1084 92
GenBank mRNA AF022818.1 - 6 - 7 838 92
AM262745.1 - 6 - 7 900 92
AM262746.1 - 6 - 7 937 92
AM262747.1 - 4 - 5 736 92
U60763.1 - 2 - 3 302 92

View Full Product Details